missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-2-Methyl-1-butanol, 98%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
100 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
100mL
2.5 L
25mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -70.0°C |
| Color | Colorless to Yellow |
| Density | 0.8100g/mL |
| Boiling Point | 130.0°C |
| Flash Point | 43°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (GC) |
| Packaging | Glass bottle |
| Linear Formula | C2H5CH(CH3)CH2OH |
| Refractive Index | 1.4090 to 1.4110 |
| Show More |
Safety and Handling
Product Identifier
- DL-2-Methyl-1-butanol
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : 2-Methyl-1-butanol
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction