missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Isopropylserine, 99%, Thermo Scientific™

Product Code. 10601642
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7522-43-2
Molecular Formula C6H13NO3
Molecular Weight (g/mol) 147.17
MDL Number MFCD01863303
InChI Key QTYSAGIKKRDGIV-ZCFIWIBFSA-N
Synonym dl-2-isopropylserine, s-2-amino-2-hydroxymethyl-3-methylbutanoic acid, 2-isopropyl-l-serine, 2s-2-amino-2-hydroxymethyl-3-methylbutanoic acid
PubChem CID 7010084
IUPAC Name (2S)-2-amino-2-(hydroxymethyl)-3-methylbutanoic acid
SMILES CC(C)C(CO)(C(=O)O)N

Specifications

Melting Point 262°C
Color Cream
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in aq. acids and bases
Formula Weight 147.17
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material DL-2-Isopropylserine, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'