missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-2-Isopropylserine, 99%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Specifications
Melting Point | 262°C |
Color | Cream |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Quantity | 1 g |
Solubility Information | Solubility in water: .. Other solubilities: soluble in aq. acids and bases |
Formula Weight | 147.17 |
Percent Purity | 99% |
Physical Form | Crystalline Powder |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'