missing translation for 'onlineSavingsMsg'
Learn More

DL-2-aminobutyric acid, TRC

Product Code. 30327717
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 30327717

Brand: Toronto Research Chemicals TRCA6143201G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DL-2-aminobutyric acid
CAS 2835-81-6
Molecular Formula C4H9NO2
Quantity 1 g
Synonym Butanoic acid, 2-amino-, Butanoic acid, 2-amino-, (±)-, Butyric acid, 2-amino-, DL- (8CI), Butyric acid, 2-amino-, dl- (5CI), Butyric acid, α-amino- (3CI), 2-Aminobutanoic acid, (±)-2-Aminobutanoic acid, (±)-2-Aminobutyric acid, (±)-α-Aminobutyric acid, 2-Amino-butyric acid, 2-Amino-n-butyric acid, 2-Aminobutyric acid, 2-Azaniumylbutanoate, AABA, Butyrine, DL-2-Amino-n-butyric acid, DL-2-Aminobutanoic acid, DL-2-Aminobutyric acid, DL-Butyrine, DL-Ethylglycine, DL-α-Amino-n-butyric acid, DL-α-Aminobutanoic acid, DL-α-Aminobutyric acid, Homoalanine, NSC 3251, dl-α-Amino-n-butyric acid, α-Amino-n-butyric acid, α-Aminobutanoic acid, α-Aminobutyric acid
InChI Formula InChI=1S/C4H9NO2/c1-2-3(5)4(6)7/h3H,2,5H2,1H3,(H,6,7)
SMILES CCC(N)C(O)=O
IUPAC Name 2-aminobutanoic acid
Molecular Weight (g/mol) 103.12
Formula Weight 103.0633
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.