missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-2-aminobutyric acid, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | DL-2-aminobutyric acid |
| CAS | 2835-81-6 |
| Molecular Formula | C4H9NO2 |
| Quantity | 500 mg |
| Synonym | Butanoic acid, 2-amino-, Butanoic acid, 2-amino-, (±)-, Butyric acid, 2-amino-, DL- (8CI), Butyric acid, 2-amino-, dl- (5CI), Butyric acid, α-amino- (3CI), 2-Aminobutanoic acid, (±)-2-Aminobutanoic acid, (±)-2-Aminobutyric acid, (±)-α-Aminobutyric acid, 2-Amino-butyric acid, 2-Amino-n-butyric acid, 2-Aminobutyric acid, 2-Azaniumylbutanoate, AABA, Butyrine, DL-2-Amino-n-butyric acid, DL-2-Aminobutanoic acid, DL-2-Aminobutyric acid, DL-Butyrine, DL-Ethylglycine, DL-α-Amino-n-butyric acid, DL-α-Aminobutanoic acid, DL-α-Aminobutyric acid, Homoalanine, NSC 3251, dl-α-Amino-n-butyric acid, α-Amino-n-butyric acid, α-Aminobutanoic acid, α-Aminobutyric acid |
| InChI Formula | InChI=1S/C4H9NO2/c1-2-3(5)4(6)7/h3H,2,5H2,1H3,(H,6,7) |
| SMILES | CCC(N)C(O)=O |
| IUPAC Name | 2-aminobutanoic acid |
| Molecular Weight (g/mol) | 103.12 |
| Formula Weight | 103.0633 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction