missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Aminobutyric acid, 99%

Product Code. 11382336
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11382336

Brand: Thermo Scientific Alfa Aesar L06035.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2835-81-6 | C4H9NO2 | 103.121 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2835-81-6
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00008093
InChI Key QWCKQJZIFLGMSD-UHFFFAOYSA-N
Synonym dl-2-aminobutyric acid, 2-aminobutyric acid, alpha-aminobutyric acid, butanoic acid, 2-amino, butyrine, dl-2-amino-n-butyric acid, h-dl-abu-oh, aaba, dl-alpha-amino-n-butyric acid, alpha-amino-n-butyric acid
PubChem CID 6657
ChEBI CHEBI:35621
IUPAC Name 2-aminobutanoic acid
SMILES CCC(C(=O)O)N

Specifications

Melting Point ∽291°C (decomposition)
Quantity 100 g
Beilstein 635889
Merck Index 14,428
Formula Weight 103.12
Percent Purity 99%
Chemical Name or Material DL-2-Aminobutyric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.