missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Amino-1,1,1-trifluoro-3-methylbutane, 97%, Thermo Scientific™

Product Code. 11955181 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11955181 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11955181 Supplier Thermo Scientific Acros Supplier No. 432570050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1582-18-9
Color Colorless to Yellow
Boiling Point 83°C to 84°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Molecular Formula C5H10F3N
Quantity 5 g
Synonym 2-methyl-1-trifluoromethyl-propylamine, dl-2-amino-3-methyl-1,1,1-trifluorobutane, dl-2-amino-1,1,1-trifluoro-3-methyl butane, d-2-amino-1,1,1-trifluoro-3-methyl butane, 2-methyl-1-trifluoromethyl propylamine, 1,1,1-trifluoro-3-methyl-2-butanamine, 1,1,1-trifluoro-3-methyl-2-butylamine, 1,1,1-trifluoro-3-methylbut-2-ylamine, 2-methyl-1-trifluoromethyl-propylamine,racemic, dl-2-amino-1,1,1-trifluoro-3-methylbutane
InChI Key YGGUXSPKQBCCRM-UHFFFAOYSA-N
SMILES CC(C)C(C(F)(F)F)N
IUPAC Name 1,1,1-trifluoro-3-methylbutan-2-amine
Molecular Weight (g/mol) 141.14
PubChem CID 44717389
Formula Weight 141.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material DL-2-Amino-1, 1, 1-trifluoro-3-methylbutane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • DL-2-Amino-1,1,1-trifluoro-3-methylbutane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.