missing translation for 'onlineSavingsMsg'
Learn More

DL-2,4-Diaminobutyric Acid Dihydrochloride, TRC

Product Code. 30384744
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384744 50 mg -
30381172 100 mg -
30386928 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384744 Supplier Toronto Research Chemicals Supplier No. TRCD45355350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65427-54-5
Molecular Formula C4H10N2O2 . 2HCl
Molecular Weight (g/mol) 191.05
Synonym Butanoic acid, 2,4-diamino-, dihydrochloride (9CI), Butanoic acid, 2,4-diamino-, dihydrochloride, (±)-, 2,4-Diaminobutanoic acid dihydrochloride, 2,4-Diaminobutyric acid dihydrochloride, NSC 41117, dl-2,4-Diaminobutyric acid dihydrochloride
SMILES Cl.Cl.NCCC(N)C(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C4H10N2O2.2ClH/c5-2-1-3(6)4(7)8;;/h3H,1-2,5-6H2,(H,7,8);2*1H
Formula Weight 190.0276
Chemical Name or Material DL-2,4-Diaminobutyric Acid Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.