missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-10-Camphorsulfonic acid, 98%, Thermo Scientific Chemicals
18.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Absorbance | (20% aq. soln.) |
Melting Point | >195°C |
Color | Beige |
Loss on Drying | 1.5% max. (105°C) |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Packaging | Plastic bottle |
Quantity | 100 g |
Beilstein | 11, 316 |
Fieser | 14,71; 15,68; 16,58 |
Show More |
Safety and Handling
Product Identifier
- DL-10-Camphorsulfonic acid
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'