missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-1,4-Dithiothreitol, For Biochemistry, 1M solution in water

Product Code. 10162994 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Unit Size:
10mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10162994 10 mL 10mL
10674545 50 mL 50mL
10215054 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10162994 Supplier Thermo Scientific Chemicals Supplier No. 426380100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3483-12-3 | C4H10O2S2 | 154.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For Biochemistry
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DL-1, 4-Dithiothreitol
Color Colorless
Name Note 1M Solution in water, For molecular biology
CAS 3483-12-3, 7732-18-5
Molecular Formula C17H20N2O6
MDL Number MFCD00053547
Concentration 0.90 to 1.10M
Density 1.0400g/mL
Merck Index 15, 3419
Synonym z-val-osu, n-cbz-l-valine succinimidyl ester, s-2,5-dioxopyrrolidin-1-yl 2-benzyloxy carbonyl amino-3-methylbutanoate, l-valine,n-phenylmethoxy carbonyl-, 2,5-dioxo-1-pyrrolidinyl ester, zvalosu, 2,5-dioxopyrrolidin-1-yl 2s-2-benzyloxy carbonyl amino-3-methylbutanoate, cbz-val-osu, ksc496o1j, benzyl s-1-2,5-dioxo-1-pyrrolidinyl oxy carbonyl-2-methylpropyl carbamate, n-cbz-valinyl oxy-succinimide
InChI Key MFAOBGXYLNLLJE-HNNXBMFYSA-N
SMILES CC(C)[C@H](NC(=O)OCC1=CC=CC=C1)C(=O)ON1C(=O)CCC1=O
IUPAC Name (2,5-dioxopyrrolidin-1-yl) (2S)-3-methyl-2-(phenylmethoxycarbonylamino)butanoate
Molecular Weight (g/mol) 348.36
PubChem CID 853556
Formula Weight 154.24
Grade Biochemical
Packaging Glass bottle
Specific Gravity 1.04
Quantity 10 mL
Physical Form Liquid
Show More Show Less
compliance-icons
Product Identifier
  • DL-Dithiothreitol, 1M solution in water
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.