missing translation for 'onlineSavingsMsg'
Learn More

DL-1-(2-Furyl)ethanol, 99%, stabilized

Product Code. 10337431 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10337431 5 g Glass bottle 5g
10736832 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10337431 Supplier Thermo Scientific Supplier No. 339670050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 4208-64-4 | C6H8O2 | 112.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4208-64-4
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.13
MDL Number MFCD00077775
InChI Key UABXUIWIFUZYQK-UHFFFAOYSA-N
Synonym 1-2-furyl ethanol, 1-furan-2-yl ethanol, 1-furan-2-yl-ethanol, 1-2-furyl ethan-1-ol, 2-furanmethanol, ?-methyl, 1-furan-2-yl ethan-1-ol, 2-furanmethanol,, a-methyl, methylfurfuryl alcohol, alpha-methylfuran-2-methanol, 1-furylethanol
PubChem CID 107243
IUPAC Name 1-(furan-2-yl)ethanol
SMILES CC(C1=CC=CO1)O

Specifications

Color Colorless to Yellow
Density 1.0700g/mL
Boiling Point 72°C to 74°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Refractive Index 1.4770 to 1.48
Quantity 5 g
Beilstein 17, V,3, 374
Specific Gravity 1.07
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in ethanol and most common organic solvent
Stabilizer 0.3 to 0.5% hydroquinone
Formula Weight 112.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material DL-1-(2-Furyl)ethanol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • DL-1-(2-Furyl)ethanol, stabilized
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.