missing translation for 'onlineSavingsMsg'
Learn More

DKM 2-93, MedChemExpress

Product Code. 16434696
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16434696 100 mg 100mg
16481536 500 mg 500mg
16400946 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16434696 Supplier Medchem Express Supplier No. HY101836
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress DKM 2-93 is a relatively selective inhibitor of UBA5 with an IC50 of 430 μM.

DKM 2-93 is a relatively selective inhibitor of UBA5 with an IC50 of 430 μM.

Specifications

Chemical Name or Material DKM 2-93
CAS 65836-72-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C11H14ClNO3
Quantity 100 mg
Solubility Information DMSO : 125 mg/mL (512.95 mM; Need ultrasonic)
SMILES O=C(NCC1=CC=C(OC)C(OC)=C1)CCl
Molecular Weight (g/mol) 243.69
Formula Weight 243.69
Percent Purity 98.07%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.