missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DKK4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113446
This item is not returnable.
View return policy
Description
DKK4 Polyclonal Antibody for Western Blot, IHC (P), ELISA
This gene encodes a protein that is a member of the dickkopf family. The secreted protein contains two cysteine rich regions and is involved in embryonic development through its interactions with the Wnt signaling pathway. Activity of this protein is modulated by binding to the Wnt co-receptor and the co-factor kremen 2.
Specifications
| DKK4 | |
| Polyclonal | |
| Unconjugated | |
| DKK4 | |
| Dickkopf gene 4; dickkopf homolog 4 (Xenopus laevis); dickkopf WNT signaling pathway inhibitor 4; dickkopf-4; Dickkopf-related protein 4; Dickkopf-related protein 4 short form; DKK4; Dkk-4; hDkk-4; MGC129562; MGC129563 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27121 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.85 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UBT3 | |
| DKK4 | |
| Recombinant Human Dickkopf-related protein 4 protein (19-224AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction