missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DKK3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10365-1-AP-20UL
This item is not returnable.
View return policy
Description
DKK3, like DKK1, DKK2, and DKK4, possesses an N-terminal signal peptide and 2 conserved cysteine-rich domains, which are separated by a linker region and contain 10 cysteine residues each. The second cysteine region has a putative lipid-binding function that may facilitate WNT/DKK interactions at the plasma membrane. The linker region contains 50 to 55 amino acids in DKK1, DKK2, and DKK4, whereas in DKK3 it contains only 12 amino acids. All DKKs have several potential sites for cleavage by furin-type proteases. Northern blot analysis revealed wide expression of the DKK3 transcript, with highest expression in heart, brain, and spinal cord. In situ hybridization reveals highest expression in mouse brain, eye, and heart.Specifications
| DKK3 | |
| Polyclonal | |
| Unconjugated | |
| DKK3 | |
| Dickkopf 3, Dickkopf related protein 3, Dkk 3, DKK3, hDkk 3, REIC, RIG | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 27122, 50781 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry, Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9QUN9, Q9UBP4 | |
| Dkk3 | |
| DKK3 Fusion Protein Ag0417 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction