missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DKK2 Monoclonal Antibody (1833CT856.7.20)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA537759
This item is not returnable.
View return policy
Description
DKK2 Monoclonal Antibody for Western Blot
This gene encodes a protein that is a member of the dickkopf family. The secreted protein contains two cysteine rich regions and is involved in embryonic development through its interactions with the Wnt signaling pathway. It can act as either an agonist or antagonist of Wnt/beta-catenin signaling, depending on the cellular context and the presence of the co-factor kremen 2. Activity of this protein is also modulated by binding to the Wnt co-receptor LDL-receptor related protein 6.
Specifications
| DKK2 | |
| Monoclonal | |
| 0.25 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9QYZ8, Q9UBU2 | |
| DKK2 | |
| recombinant protein of human DKK2. | |
| 200 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 κ |
| Western Blot | |
| 1833CT856.7.20 | |
| Unconjugated | |
| DKK2 | |
| dickkopf 2; dickkopf 2 homolog; Dickkopf gene 2; dickkopf homolog 1; dickkopf homolog 2; dickkopf homolog 2 (Xenopus laevis); dickkopf homolog 4; dickkopf related protein-2; dickkopf WNT signaling pathway inhibitor 2; dickkopf-2; Dickkopf-related protein 2; DKK2; DKK-2; Dkk4; hDkk-2; mDkk-2; UNQ682/PRO1316 | |
| Mouse | |
| Protein G | |
| RUO | |
| 27123, 56811 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction