missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DKK1 Recombinant Human Monoclonal Antibody (27H11)
GREENER_CHOICE

Human Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA560724

Product Code. 30535038

  • £382.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

DKK1 Recombinant Monoclonal Antibody for ELISA

DKK-1 is a member of the DKK protein family which also includes DKK-2, DKK-3 and DKK-4. DKK-1 was originally identified as a Xenopus head-forming molecule that behaves as an antagonist for Wnt signaling. Subsequent studies have shown that DKK-1 and DKK-4 play important regulatory roles in the Wnt/beta-catenin signaling pathway by forming inhibitory complexes with LDL receptor-related proteins 5 and 6 (LRP5 and LRP6), which are essential components of the Wnt/beta-catenin signaling system. LRP5 and LRP6 are single-pass transmembrane proteins that appear to act as co-receptors for Wnt ligands involved in the Wnt/beta-catenin signaling cascade. It has been suggested that by inhibiting Wnt/beta-catenin signaling, which is essential for posterior patterning in vertebrates, DKK-1 permits anterior development. This notion is supported by the finding that mice deficient of DKK-1 expression lack head formation and die during embryogenesis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DKK1
Recombinant Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
O94907
DKK1
Recombinant Human DKK1 protein.
100 μL
Primary
Human
Antibody
IgG1
ELISA
27H11
Unconjugated
DKK1
dickkopf 1 homolog; dickkopf homolog 1; dickkopf homolog 1 (Xenopus laevis); dickkopf related protein-1; dickkopf WNT signaling pathway inhibitor 1; Dickkopf1; dickkopf-1; dickkopf-1 like; dickkopf-like protein 1; dickkopf-related protein 1; DKK1; DKK-1; hDkk1; hDkk-1; mdkk-1; SK; UNQ492/PRO1008
Human
Affinity chromatography
RUO
22943
-20°C or -80°C if preferred
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.