Learn More
Invitrogen™ DKK1 Mouse ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EMDKK1X5
Additional Details : Weight : 0.33800kg
Description
The Mouse Dkk-1 ELISA quantitates Ms Dkk-1 in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms Dkk-1. Principle of the method The Mouse Dkk-1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
This gene encodes a protein that is a member of the dickkopf family. It is a secreted protein with two cysteine rich regions and is involved in embryonic development through its inhibition of the WNT signaling pathway. Elevated levels of DKK1 in bone marrow plasma and peripheral blood is associated with the presence of osteolytic bone lesions in patients with multiple myeloma.Specifications
O54908 | |
25 pg/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
5 x 96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
13380 | |
<12% | |
Chromogen, Biotinylated Detection Antibody, Pre-coated 96 well plates, Assay Diluent Concentrate, Stop Solution, Standard, Adhesive Plate Covers, Wash Buffer, SAV-HRP | |
5 x 96 Tests | |
Development | |
2°C to 8°C | |
Mouse | |
4 hr. 45 min. |
24.58 pg/mL to 6000 pg/mL | |
25 pg/mL to 6000 pg/mL | |
Biotin | |
Other Proteins | |
This ELISA kit shows no cross-reactivity with the following cytokines tested: Mouse CD30L, CD30, CD40, CRG-2, CTACK, CXCL16, Eotaxin, Eotaxin-2, Fas Ligand, Fractalkine, GCSF, GM-CFS, IFN- gamma, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-3 Rb, IL- 4, IL-5, IL-9, IL-10, IL-12 p40/p70, IL-12 p70, IL-13, IL-17, KC, Leptin R, LEPTIN(OB), LIX, L-Selectin, Lymphotactin, MCP-1, MCP-5, M-CSF, MIG, MIP-1 alpha, MIP-1 gamma, MIP-2, MIP-3 beta, MIP-3 alpha, PF-4, P-Selectin, RANTES, SCF, SDF-1 alpha, TARC, TCA-3, TECK, TIMP-1, TNF-alpha, TNF RI, TNF RII, TPO, VCAM-1, VEGF. | |
Mouse | |
Colorimetric Microplate Reader | |
Dkk1 | |
<10% | |
HRP | |
RUO | |
Plasma, 50 μ;L; Serum, 50 μ;L; Supernatant, 100 μ;L | |
mdkk-1 | |
1 hr. 20 min. |