Learn More
Invitrogen™ DKK1 Monoclonal Antibody (2H2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515497
Description
MA5-15497 targets DKK1 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15497 immunogen is purified recombinant fragment of DKK1 expressed in E. Coli. MA5-15497 detects DKK1 which has a predicted molecular weight of approximately 28.7kDa.
DKK-1 is a member of the DKK protein family which also includes DKK-2, DKK-3 and DKK-4. DKK-1 was originally identified as a Xenopus head-forming molecule that behaves as an antagonist for Wnt signaling. Subsequent studies have shown that DKK-1 and DKK-4 play important regulatory roles in the Wnt/beta-catenin signaling pathway by forming inhibitory complexes with LDL receptor-related proteins 5 and 6 (LRP5 and LRP6), which are essential components of the Wnt/beta-catenin signaling system. LRP5 and LRP6 are single-pass transmembrane proteins that appear to act as co-receptors for Wnt ligands involved in the Wnt/beta-catenin signaling cascade. It has been suggested that by inhibiting Wnt/beta-catenin signaling, which is essential for posterior patterning in vertebrates, DKK-1 permits anterior development. This notion is supported by the finding that mice deficient of DKK-1 expression lack head formation and die during embryogenesis.
Specifications
| DKK1 | |
| Monoclonal | |
| Conc. Not Determined | |
| ascites with 0.03% sodium azide | |
| O94907 | |
| DKK1 | |
| Purified recombinant fragment of DKK1 expressed in E. Coli. | |
| RUO | |
| 22943 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| 2H2 | |
| Unconjugated | |
| DKK1 | |
| dickkopf 1 homolog; dickkopf homolog 1; dickkopf homolog 1 (Xenopus laevis); dickkopf related protein-1; dickkopf WNT signaling pathway inhibitor 1; Dickkopf1; dickkopf-1; dickkopf-1 like; dickkopf-like protein 1; dickkopf-related protein 1; DKK1; DKK-1; hDkk1; hDkk-1; mdkk-1; SK; UNQ492/PRO1008 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.