missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dkk-3 Rabbit anti-Human, Mouse, Rat, Clone: 1R5E0, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | Dkk-3 |
|---|---|
| Clone | 1R5E0 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18362137
|
Novus Biologicals
NBP3-16293-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18371064
|
Novus Biologicals
NBP3-16293-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dkk-3 Monoclonal antibody specifically detects Dkk-3 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| Dkk-3 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 27122 | |
| IgG | |
| Affinity purified |
| 1R5E0 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| dickkopf (Xenopus laevis) homolog 3, dickkopf 3, dickkopf homolog 3 (Xenopus laevis), Dickkopf-3, dickkopf-related protein 3, Dkk-3, hDkk-3, regulated in glioma, REICdkk-3, RIG, RIG-like 5-6, RIG-like 7-1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 200-360 of human Dkk-3 (Q9UBP4). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title