missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dkk-3 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-899
This item is not returnable.
View return policy
Description
Dkk-3 Polyclonal specifically detects Dkk-3 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| Dkk-3 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| dickkopf (Xenopus laevis) homolog 3, dickkopf 3, dickkopf homolog 3 (Xenopus laevis), Dickkopf-3, dickkopf-related protein 3, Dkk-3, hDkk-3, regulated in glioma, REICdkk-3, RIG, RIG-like 5-6, RIG-like 7-1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| NP_037385.2; NP_056965.3 and NP_001018067.1 are varients of the same protein. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/mL, Peptide ELISA Detection limit 1:16000 | |
| NP_037385.2 | |
| DKK3 | |
| Peptide with sequence C-AAAALLGGEEI corresponding to C-Terminus according to NP_037385.2, NP_056965.3, NP_001018067.1. | |
| 0.1 mg | |
| Signal Transduction | |
| 27122 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction