missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Dkk-3 Monoclonal antibody specifically detects Dkk-3 in Human samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | Dkk-3 |
| Conjugate | mFluor Violet 610 SE |
| Quantity | 0.1 mL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 27122 |
| Isotype | IgG1 κ |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?