missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dkk-3 Antibody (103B5), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-60242AF647
This item is not returnable.
View return policy
Description
Dkk-3 Monoclonal antibody specifically detects Dkk-3 in Human samples. It is validated for Western Blot, ELISA
Specifications
| Dkk-3 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| dickkopf (Xenopus laevis) homolog 3, dickkopf 3, dickkopf homolog 3 (Xenopus laevis), Dickkopf-3, dickkopf-related protein 3, Dkk-3, hDkk-3, regulated in glioma, REICdkk-3, RIG, RIG-like 5-6, RIG-like 7-1 | |
| Recombinant human DKK-3 (22-350aa) purified from E. coli | |
| 0.1 mL | |
| Signal Transduction | |
| 27122 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA | |
| 103B5 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction