missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dkk-3 Antibody (019), Alexa Fluor™ 532, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90496AF532
This item is not returnable.
View return policy
Description
Dkk-3 Monoclonal specifically detects Dkk-3 in Mouse samples. It is validated for ELISA.
Specifications
| Dkk-3 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| dickkopf (Xenopus laevis) homolog 3, dickkopf 3, dickkopf homolog 3 (Xenopus laevis), Dickkopf-3, dickkopf-related protein 3, Dkk-3, hDkk-3, regulated in glioma, REICdkk-3, RIG, RIG-like 5-6, RIG-like 7-1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Dkk-3 (NP_056629.1; Met 1-Ile 349). | |
| 0.1 mL | |
| Signal Transduction | |
| 27122 | |
| Mouse | |
| Purified |
| ELISA | |
| 19 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| No cross-reactivity in ELISA with: Human DKK3 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction