Learn More
Dkk-2 Antibody (994930), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6628MFV610
This item is not returnable.
View return policy
Description
Dkk-2 Monoclonal antibody specifically detects Dkk-2 in Human samples. It is validated for Immunohistochemistry, Flow Cytometry, Immunocytochemistry
Specifications
| Dkk-2 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Stem Cells | |
| 27123 | |
| Store at 4C in the dark. |
| Immunohistochemistry, Flow Cytometry, Immunocytochemistry | |
| 994930 | |
| Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry | |
| dickkopf (Xenopus laevis) homolog 2, dickkopf 2, dickkopf homolog 2 (Xenopus laevis), dickkopf related protein-2, dickkopf-2, dickkopf-related protein 2, DKK-2, hDkk-2 | |
| Chinese Hamster Ovary cell line, CHO-derived human Dkk-2, Met1-Ile259, Accession # NP_055236 | |
| RUO | |
| Primary | |
| Human | |
| IgG2a |