missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DKFZP434B168 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92294-0.1ml
This item is not returnable.
View return policy
Description
DKFZP434B168 Polyclonal antibody specifically detects DKFZP434B168 in Human, Mouse samples. It is validated for Western Blot
Specifications
| DKFZP434B168 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| C1orf73, chromosome 1 open reading frame 73, DKFZp434B168, Int7, integrator complex subunit 7 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 702-948 of human INTS7 (NP_001186740.1). NVELQQQSCLLISHAIEALILDPESASFQEYGSTGTAHADSEYERRMMSVYNHVLEEHTACLCNAIIALLKVPLSFQRYFFQKLQSTSIKLALSPSPRNPAEPIAVQNNQQLALKVEGVVQHGSKPGLFRKIQSVCLNVSSTLQSKSGQDYKIPIDNMTNEMEQRVEPHNDYFSTQFLLNFAILGTHNITVESSVKDANGIVWKTGPRTTIFVKSLEDPYSQQIRLQQQQAQQPLQQQQQRNAYTRF | |
| 0.1 mL | |
| Zinc Finger | |
| 25896 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction