missing translation for 'onlineSavingsMsg'
Learn More

DKFZP434B168 Antibody [Alexa Fluor™ 647], Novus Biologicals™

Product Code. 30610635 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30610635 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30610635 Supplier Novus Biologicals Supplier No. NBP346470AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DKFZP434B168 Polyclonal antibody specifically detects DKFZP434B168 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen DKFZP434B168
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias C1orf73, chromosome 1 open reading frame 73, DKFZp434B168, Int7, integrator complex subunit 7
Host Species Rabbit
Immunogen integrator complex subunit 7. (Uniprot# Q9NVH2)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 25896
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.