missing translation for 'onlineSavingsMsg'
Learn More

DKC1 Rabbit anti-Human, Mouse, Rat, Clone: 2W2V6, Novus Biologicals™

Product Code. 18321657
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18321657 20 μg 20µL
18378583 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18321657 Supplier Novus Biologicals Supplier No. NBP31640520UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

DKC1 Monoclonal antibody specifically detects DKC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen DKC1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2W2V6
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias CBF5, CBF5 homolog, cbf5p homolog, DKC, dyskeratosis congenita 1, dyskerin, Dyskerin, EC 5.4.99, EC 5.4.99.-, FLJ97620, H/ACA ribonucleoprotein complex subunit 4, NAP57, NOLA4dyskerin, Nopp140-associated protein of 57 kDa, Nucleolar protein family A member 4, Nucleolar protein NAP57, snoRNP protein DKC1, XAP101
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 300-400 of human DKC1 (O60832).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1736
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.