missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DJ-1 Recombinant Rabbit Monoclonal Antibody (23GB1955), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553182
This item is not returnable.
View return policy
Description
The product of this gene belongs to the peptidase C56 family of proteins. It acts as a positive regulator of androgen receptor-dependent transcription. It may also function as a redox-sensitive chaperone, as a sensor for oxidative stress, and it apparently protects neurons against oxidative stress and cell death. Defects in this gene are the cause of autosomal recessive early-onset Parkinson disease 7. Two transcript variants encoding the same protein have been identified for this gene.
Specifications
| DJ-1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Park7 | |
| CAP1; Contraception-associated protein 1; CTA-215D11.1; dj1; DJ-1; DJ-1 protein; early onset 7; epididymis secretory sperm binding protein Li 67p; fertility protein SP22; FLJ27376; FLJ34360; FLJ92274; HEL-S-67p; hypothetical protein LOC449674; maillard deglycase; Oncogene DJ1; OTTHUMP00000001350; OTTHUMP00000001351; PARK7; PARK7 GENE; PARK7/DJ1; parkinson disease (autosomal recessive, early onset) 7; Parkinson disease 7; Parkinson disease protein 7; Parkinson disease protein 7 homolog; Parkinson disease protein 7 homolog; protein/nucleic acid deglycase DJ-1; parkinson protein 7; Parkinsonism associated deglycase; parkinsonism-associated deglycase; Protein CAP1; protein deglycase DJ-1; protein deglycase DJ-1zDJ-1; Protein DJ-1; protein DJ-1-like protein; protein DJ-1zDJ-1; Protein/nucleic acid deglycase DJ-1; RNA-binding protein regulatory subunit; SP22; zDJ-1; zgc:103725 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11315, 117287, 57320 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB1955 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O88767, Q99497, Q99LX0 | |
| Park7 | |
| A synthesized peptide derived from human PARK7 (1-66AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction