missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DJ-1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518140
This item is not returnable.
View return policy
Description
This antibody is predicted to react with mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
The product of this gene belongs to the peptidase C56 family of proteins. It acts as a positive regulator of androgen receptor-dependent transcription. It may also function as a redox-sensitive chaperone, as a sensor for oxidative stress, and it apparently protects neurons against oxidative stress and cell death. Defects in this gene are the cause of autosomal recessive early-onset Parkinson disease 7. Two transcript variants encoding the same protein have been identified for this gene.
Specifications
| DJ-1 | |
| Polyclonal | |
| Unconjugated | |
| Park7 | |
| CAP1; Contraception-associated protein 1; CTA-215D11.1; dj1; DJ-1; DJ-1 protein; early onset 7; epididymis secretory sperm binding protein Li 67p; fertility protein SP22; FLJ27376; FLJ34360; FLJ92274; HEL-S-67p; hypothetical protein LOC449674; maillard deglycase; Oncogene DJ1; OTTHUMP00000001350; OTTHUMP00000001351; PARK7; PARK7 GENE; PARK7/DJ1; parkinson disease (autosomal recessive, early onset) 7; Parkinson disease 7; Parkinson disease protein 7; Parkinson disease protein 7 homolog; Parkinson disease protein 7 homolog; protein/nucleic acid deglycase DJ-1; parkinson protein 7; Parkinsonism associated deglycase; parkinsonism-associated deglycase; Protein CAP1; protein deglycase DJ-1; protein deglycase DJ-1zDJ-1; Protein DJ-1; protein DJ-1-like protein; protein DJ-1zDJ-1; Protein/nucleic acid deglycase DJ-1; RNA-binding protein regulatory subunit; SP22; zDJ-1; zgc:103725 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 11315, 117287, 57320 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O88767, Q99497, Q99LX0 | |
| Park7 | |
| Synthetic peptide sequence (AAQVKAPLVLKD) corresponding to the C-terminus amino acids of PARK7 (aa 178-189). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction