missing translation for 'onlineSavingsMsg'
Learn More

(-)-Dizocilpine maleate, MedChemExpress

Product Code. 16486308
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486308 10 mg 10mg
16462427 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16486308 Supplier Medchem Express Supplier No. HY15084A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (-)-Dizocilpine maleate ((-)-MK-801 maleate) is a less active (-)-enantiomer of Dizocilpine. (-)-Dizocilpine maleate is a selective and non-competitive N-methyl-D-aspartate (NMDA) receptor antagonist with a Ki of 211.7 nM. (-)-Dizocilpine maleate has antidepressant effects.

(-)-Dizocilpine maleate ((-)-MK-801 maleate) is a less active (-)-enantiomer of Dizocilpine. (-)-Dizocilpine maleate is a selective and non-competitive N-methyl-D-aspartate (NMDA) receptor antagonist with a Ki of 211.7 nM. (-)-Dizocilpine maleate has antidepressant effects.

Specifications

Chemical Name or Material (-)-Dizocilpine maleate
CAS 121917-57-5
Color White
Purity Grade Notes Research
Molecular Formula C20H19NO4
Quantity 10 mg
Synonym (-)-MK-801 maleate
Solubility Information DMSO : 100 mg/mL (296.41 mM; Need ultrasonic) ∣Ethanol : 25 mg/mL (74.10 mM; Need ultrasonic) ∣H2O : 8.33 mg/mL (24.69 mM; ultrasonic and warming and heat to 60°C)
SMILES C[C@@]1(N2)C3=CC=CC=C3C[C@H]2C4=CC=CC=C14.O=C(O)/C=C\C(O)=O
Molecular Weight (g/mol) 337.37
Formula Weight 337.37
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.