missing translation for 'onlineSavingsMsg'
Learn More

Dixanthogen, MedChemExpress

Product Code. 16464567
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16464567 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16464567 Supplier Medchem Express Supplier No. HYB1186

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dixanthogen is an ectoparasiticide.

Dixanthogen is an ectoparasiticide.

Specifications

Chemical Name or Material Dixanthogen
Physical Form Oil
CAS 502-55-6
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C6H10O2S4
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 250 mg/mL (1031.35 mM; Need ultrasonic)
SMILES CCOC(SSC(OCC)=S)=S
Molecular Weight (g/mol) 242.4
Formula Weight 242.4
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.