missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Divozilimab Recombinant Monoclonal Antibody

Recombinant Monoclonal Antibody
£397.00 - £1002.00
Specifications
| Antigen | Divozilimab |
|---|---|
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30283485
![]() |
Invitrogen™
MA559225 |
100 μg |
£397.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30283869
|
Invitrogen™
MA559226 |
1 mg |
£1002.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Divozilimab, an anti-CD20 monoclonal antibody, is developed for the treatment of multiple sclerosis, specifically relapsing-remitting multiple sclerosis (RRMS) and secondary progressive MS (SPMS) with relapses. The CD20 gene, also known as MS4A1, encodes a B cell-specific membrane-bound protein that plays a crucial role in B cell development and function. Located on chromosome 11q12 in humans, the MS4A1 gene spans 16 kb and includes eight exons. CD20 is a part of the membrane-spanning 4-domain family, subfamily A (MS4A), which encompasses 18 tetraspan proteins. The prominent CD20 mRNA variant is 2.8 kb long, although other shorter variants exist. This phosphoprotein, approximately 33-37 kDa, is expressed on the surface of almost all normal and malignant B cells, making it an ideal target for therapeutic antibodies like rituximab. CD20 is known to interact with several proteins such as CD40, MHCII, and CD81, playing a role in B cell receptor signaling and other immune processes. Despite its widespread clinical use in B cell depletion therapies, the precise biological function of CD20 remains partly unresolved.Specifications
| Divozilimab | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Lyophilized | |
| IgG1 | |
| Human | |
| Primary | |
| Protein A |
| 1 mg/mL | |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| Unconjugated | |
| Human | |
| RUO | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title