missing translation for 'onlineSavingsMsg'
Learn More

Divinyltin dichloride, Thermo Scientific™

Product Code. 11354547
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354547 10 g 10g
11374547 2 g 2g
11364547 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11354547 Supplier Thermo Scientific Alfa Aesar Supplier No. 039613.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7532-85-6
Molecular Formula C4H8Cl2Sn
Molecular Weight (g/mol) 245.72
MDL Number MFCD00058728
InChI Key OFJXSWMLVIFLKC-UHFFFAOYSA-N
Synonym divinyltin dichloride, dichlorodivinylstannane, stannane, dichlorodiethenyl, dichloro divinyl stannane, divinyltindichloride, acmc-20al40, dichloro-bis ethenyl stannane
PubChem CID 82033
IUPAC Name dichloro-bis(ethenyl)stannane
SMILES Cl.Cl.C=C[SnH2]C=C

Specifications

Boiling Point 54°C to 56°C (3 mmHg)
Quantity 10 g
UN Number UN2788
Sensitivity Moisture Sensitive
Formula Weight 243.69
Physical Form Liquid
Chemical Name or Material Divinyltin dichloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.