missing translation for 'onlineSavingsMsg'
Learn More

Divinyltetramethyldisiloxane, 97%

Product Code. 10239873 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10239873 250 g Glass bottle 250g
10736592 10 g Glass bottle 10g
10693342 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10239873 Supplier Thermo Scientific Acros Supplier No. 249212500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2627-95-4 | C8H18OSi2 | 186.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2627-95-4
Molecular Formula C8H18OSi2
Molecular Weight (g/mol) 186.40
MDL Number MFCD00014933
InChI Key BITPLIXHRASDQB-UHFFFAOYSA-N
Synonym 1,3-divinyltetramethyldisiloxane, 1,1,3,3-tetramethyl-1,3-divinyldisiloxane, disiloxane, 1,3-diethenyl-1,1,3,3-tetramethyl, 1,1'-divinyltetramethyldisiloxane, unii-sj4d71zqf4, bisvinyltetramethyldisiloxane, 1,3-divinyl-1,1,3,3-tetramethyl disiloxane, sym-tetramethyldivinyldisiloxane, tetramethyl-1,3-divinyldisiloxane, 1,3-diethenyl-1,1,3,3-tetramethyldisiloxane
PubChem CID 75819
IUPAC Name ethenyl-[ethenyl(dimethyl)silyl]oxy-dimethylsilane
SMILES C[Si](C)(O[Si](C)(C)C=C)C=C

Specifications

Melting Point -99.7°C
Color Colorless
Density 0.8130g/mL
Boiling Point 139°C
Flash Point 24°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula [H2C=CHSi(CH3)2]2O
Refractive Index 1.4110 to 1.413
Quantity 250 g
Specific Gravity 0.813
Solubility Information Solubility in water: insoluble
Formula Weight 186.4
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Divinyltetramethyldisiloxane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Divinyltetramethyldisiloxane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.