missing translation for 'onlineSavingsMsg'
Learn More

Divinylbenzene, 80%, mixture of isomers, stab. with 1000ppm 4-tert-butylcatechol

Product Code. 11379977
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11379977 500 g 500g
11369977 25 g 25g
11359977 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11379977 Supplier Thermo Scientific Alfa Aesar Supplier No. 042804.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1321-74-0 | C10H10 | 130.19 g/mol

Divinylbenzene is used as a reactive monomer in polyester resins and for the synthesis of highly crosslinked macroporous styrene as well as a crosslinking agent. It acts as an intermediate in the production of ion exchange resin. Further, it is used in making rubber, petroleum production and polymeric core shell nanoparticles. In addition, it is used to synthesize monodispersed poly divinylbenzene microspheres.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1321-74-0
Molecular Formula C10H10
Molecular Weight (g/mol) 130.19
MDL Number MFCD01778656,MFCD00010654
InChI Key MYRTYDVEIRVNKP-UHFFFAOYSA-N
Synonym divinylbenzene, 1,2-divinylbenzene, o-divinylbenzene, benzene, diethenyl, benzene, 1,2-diethenyl, divinylbenzen, divinyl benzen, divinyl-benzene, poly divinylbenzene, divinylbenzene dvb
PubChem CID 66666
IUPAC Name 1,2-bis(ethenyl)benzene
SMILES C=CC1=CC=CC=C1C=C

Specifications

Color Colorless to Yellow
Density 0.914 g/mL
Flash Point 69°C (156°F)
Refractive Index 1.574
Quantity 500 g
Sensitivity Light sensitive
Solubility Information Miscible with ethanol and ether. Immiscible with water.
Formula Weight 130.19
Percent Purity 80%
Physical Form Liquid
Chemical Name or Material Divinylbenzene, Mixture of isomers, Stabilized with 1000ppm 4-tert-butylcatechol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Divinylbenzene, 80%, mixture of isomers, stabilized with 1000ppm 4-tert-butylcatechol
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: divinylbenzene, (1-methylenepropyl)-benzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.