missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Divinyl Sulfone (Stabilized with Hydroquinone), TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | Divinyl Sulfone |
| CAS | 77-77-0 |
| Molecular Formula | C4H6O2S |
| Quantity | 50 mg |
| Synonym | DivinylSulfone |
| InChI Formula | InChI=1S/C4H6O2S/c1-3-7(5,6)4-2/h3-4H,1-2H2 |
| SMILES | C=CS(=O)(=O)C=C |
| IUPAC Name | 1-ethenylsulfonylethene |
| Molecular Weight (g/mol) | 118.16 |
| Formula Weight | 118.0089 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction