Learn More
BINDER™ Divided Inner Glass Door for Series CB, CBF
Product Code. 17373683 Shop All BINDER Products
Click to view available options
Door Style:
4-part Internal Door
6-part Internal Door
8-part Internal Door
Unit Size:
Each
Product Code. 17373683
Brand: BINDER™ undefined
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
BINDER™ Divided inner glass door is an option for use with CO2 incubators CB and CBF
  • Inner door divided and compartmentation
  • Internal door divided into 4, 6 or 8 (depending on model) parts and compartmentation of the interior
Specifications
Description Inner door, divided - Cell therapy compartmentalization, consisting of 4 inner doors and two shelf levels with one partition wall each
For Use With (Equipment) BINDER Series CB and CBF
Material Silicone
Type Access Port
Door Style 4-part Internal Door
Features Internal door divided into 4 parts