missing translation for 'onlineSavingsMsg'
Learn More

Diuron, TRC

Product Code. 30381648
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30381648 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30381648 Supplier Toronto Research Chemicals Supplier No. TRCD4944151G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diuron
CAS 330-54-1
Molecular Formula C9 H10 Cl2 N2 O
Quantity 1 g
Synonym Urea, N'-(3,4-dichlorophenyl)-N,N-dimethyl-, Urea, 3-(3,4-dichlorophenyl)-1,1-dimethyl- (8CI), N'-(3,4-Dichlorophenyl)-N,N-dimethylurea, 1,1-Dimethyl-3-(3,4-dichlorophenyl)urea, 1-(3,4-Dichlorophenyl)-3,3-dimethylurea, 101S, 3-(3,4-Dichlorophenyl)-1,1-dimethylurea, Biocut DC 100, Cardi, DCMC, DCMU, DCMU 9, DCMU 99, DMU, DP Hardener 95, Dairon, Direx, Dironet, Dironzol, Diurex, Diuron, Diuron Nortox, Duran, Dyhard 200, Dyhard UR 200, HRT Dinron, HW 920, Herbatox, Karmax, Karmex, Karmex D, Karmex DW, Karmex Diuron Herbicide, Lucenit, Marmer, N,N-Dimethyl-N'-(3,4-dichlorophenyl)urea, N-(3,4-Dichlorophenyl)-N',N'-dimethylurea, NSC 8950, Preventol A 6, SM 400, SM 400 (curing agent), Telvar Diuron Weed Killer, UR 200
InChI Formula InChI=1S/C9H10Cl2N2O/c1-13(2)9(14)12-6-3-4-7(10)8(11)5-6/h3-5H,1-2H3,(H,12,14)
SMILES CN(C)C(=O)Nc1ccc(Cl)c(Cl)c1
IUPAC Name 3-(3,4-dichlorophenyl)-1,1-dimethylurea
Molecular Weight (g/mol) 233.09
Formula Weight 232.017
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.