missing translation for 'onlineSavingsMsg'
Learn More

Ditridecyl Adipate, TRC

Product Code. 30135361
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135361 2.5 g 2.5g
30011461 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135361 Supplier Toronto Research Chemicals Supplier No. TRCD4941952.5G
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16958-92-2
Molecular Formula C32H62O4
Molecular Weight (g/mol) 510.84
Synonym Hexanedioic Acid 1,6-Ditridecyl Ester, Adipic Acid Ditridecyl Ester, Hexanedioic Acid Ditridecyl Ester, 1-Tridecanol Adipate (2:1), Bis(tridecyl) Adipate, Cereplas DTDA, Emkarate DTDA, Esterex A 51, Plasthall DTDA
IUPAC Name ditridecyl adipate
SMILES O=C(CCCCC(OCCCCCCCCCCCCC)=O)OCCCCCCCCCCCCC

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C32H62O4/c1-3-5-7-9-11-13-15-17-19-21-25-29-35-31(33)27-23-24-28-32(34)36-30-26-22-20-18-16-14-12-10-8-6-4-2/h3-30H2,1-2H3
Formula Weight 510.46
Chemical Name or Material Adipic Acid-bistridecyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.