missing translation for 'onlineSavingsMsg'
Learn More

Dithranol, Mikromol™

Product Code. 17911286
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17911286 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17911286 Supplier Mikromol Supplier No. MM0137.00
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Dithranol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dithranol
CAS 1143-38-0
Analyte or Component Names Dithranol
Shipping Condition Room Temperature
Molecular Formula C14 H10 O3
Certifications/Compliance ISO 17034
Synonym 1,8-Dihydroxy-9(10H)-anthracenone, 1,8-Dihydroxyanthrone, 1,8-Dihydroxy-9-anthrone, 1,8-Dihydroxyanthrone, Anthra-Derm, Anthralin, Batidrol, Batridol, Chrysodermol, Cignolin, Cigthranol, Derobin, Dithranol, DrithoCreme, Drithoscalp, Lasan, NSC 43970, NSC 629313, Psodadrate, Psoriacid-Stift, Psoriacide
InChI Formula InChI=1S/C14H10O3/c15-10-5-1-3-8-7-9-4-2-6-11(16)13(9)14(17)12(8)10/h1-6,15-16H,7H2
SMILES Oc1cccc2Cc3cccc(O)c3C(=O)c12
IUPAC Name 1,8-dihydroxy-10H-anthracen-9-one
Molecular Weight (g/mol) 226.23
Quantity 500 mg
Formula Weight 226.063 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.