missing translation for 'onlineSavingsMsg'
Learn More

Dithranol EP Impurity C, Mikromol™

Product Code. 30050952
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050952 25 mg 25mg
17933566 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050952 Supplier Mikromol Supplier No. MM0137.020025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Dithranol EP Impurity C
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,4',5,5'-Tetrahydroxy-9,9'-bianthracenyl-10,10'(9H,9'H)-dione
CAS 31991-54-5
Analyte or Component Names 4,4',5,5'-Tetrahydroxy-9,9'-bianthracenyl-10,10'(9H,9'H)-dione
Shipping Condition Room Temperature
Molecular Formula C28 H18 O6
Certifications/Compliance ISO 9001
Synonym 4,4',5,5'-Tetrahydroxy-9,9'-bianthracenyl-10,10'(9H,9'H)-dione (Dithranol Dimer), 4,4',5,5'-Tetrahydroxy-9,9'-bianthracenyl-10,10'(9H,9'H)-dione, Dithranol Imp. C (EP), Dithranol Dimer, Dithranol Impurity C
InChI Formula InChI=1S/C28H18O6/c29-17-9-1-5-13-21(14-6-2-10-18(30)24(14)27(33)23(13)17)22-15-7-3-11-19(31)25(15)28(34)26-16(22)8-4-12-20(26)32/h1-12,21-22,29-32H
SMILES Oc1cccc2C(C3c4cccc(O)c4C(=O)c5c(O)cccc35)c6cccc(O)c6C(=O)c12
IUPAC Name 10-(4,5-dihydroxy-10-oxo-9H-anthracen-9-yl)-1,8-dihydroxy-10H-anthracen-9-one
Molecular Weight (g/mol) 450.44
Quantity 25 mg
Formula Weight 450.1103 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.