missing translation for 'onlineSavingsMsg'
Learn More

Dithranol, 99%, MP Biomedicals™

Product Code. 11414742 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
0.25 g
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414742 0.25 g 250mg
11424742 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11414742 Supplier MP Biomedicals Supplier No. 0219150283
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Dithranol ≥99%, Yellow Powder

  • Dithranol, a potential anti-psoriasis medication, may be used to study its safety, efficacy, metabolism and pharmacological profile.
  • It may be used to study is physicochemical properties, as a reference compound and to develop effective drug delivery vehicles.
  • It may be used as a matrix for matrix assisted laser desorption/ionization imaging on a Fourier transform ion cyclotron resonance mass spectrometer.
  • Anthralin is a leukotriene biosynthesis inhibitor.
  • Inhibits LTB4 ω-oxidation and disrupts mitochondria function.
  • Anthralin is used in the treatment of psoriasis, as a fungicide, in the treatment of ringworm infections and in chronic dermatoses.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1143-38-0
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.23
MDL Number MFCD00053409
InChI Key NUZWLKWWNNJHPT-UHFFFAOYSA-N
PubChem CID 2202
ChEBI CHEBI:37510
IUPAC Name 1,8-dihydroxy-9,10-dihydroanthracen-9-one
SMILES OC1=CC=CC2=C1C(=O)C1=C(O)C=CC=C1C2

Specifications

Color Orange-Yellow or Yellow
Assay Percent Range ≥99%
Quantity 0.25 g
Synonym anthralin, dithranol, 1,8-dihydroxyanthrone, 1,8-dihydroxy-9-anthrone, chrysodermol, cigthranol, batridol, cignolin, psoriacid-stift, 9 10h-anthracenone, 1,8-dihydroxy
Formula Weight 226.23
Physical Form Crystalline Powder
Chemical Name or Material Dithranol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.