missing translation for 'onlineSavingsMsg'
Learn More

Dithizone, ACS, 85+%

Product Code. 11387588
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387588 10 g 10g
11397588 2 g 2g
11307598 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11387588 Supplier Thermo Scientific Alfa Aesar Supplier No. 033283.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 60-10-6 | C13H12N4S | 256.33 g/mol

Dithizone is used as an indicator for minute amounts of lead. It acts as a chelating agent for heavy metal poisoning. As a ligand, it forms complexes with metals like zinc, lead and mercury. Further, it is used to assess the purity of human pancreatic islet preparations, which is used for transplantation in patients with type 1 diabetes. In addition to this, it is used as a titrant in the quantitative determination of heavy metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-10-6
Molecular Formula C13H12N4S
Molecular Weight (g/mol) 256.33
MDL Number MFCD00003025
InChI Key UOFGSWVZMUXXIY-UHFFFAOYSA-N
Synonym dithizone, dithizon, ditizon, carbazone, diphenylthio, usaf ek-3092, diazenecarbothioic acid, phenyl-, 2-phenylhydrazide, 1,5-diphenyl-3-mercaptoformazan, 3-formazanthiol, 1,5-diphenyl, unii-njz2cj4d6p, 1,5-diphenyl-3-thiocarbazone
PubChem CID 657262
IUPAC Name 1-anilino-3-phenyliminothiourea
SMILES S=C(NNC1=CC=CC=C1)N=NC1=CC=CC=C1

Specifications

Melting Point 168°C (decomposition)
Odor Odorless
Quantity 10 g
Beilstein 748838
Merck Index 14,3377
Solubility Information Soluble in ethanol,chloroform,dimethyl sulfoxide and pyridine. Insoluble in water.
Formula Weight 256.33
Percent Purity ≥85%
Grade ACS
Physical Form Crystalline
Chemical Name or Material Dithizone
Show More Show Less
compliance-icons
Product Identifier
  • Dithizone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Dithizone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.