missing translation for 'onlineSavingsMsg'
Learn More

Dithizone, ACS, 85+%, Thermo Scientific Chemicals

Product Code. 11397588
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Dithizone is used as an indicator for minute amounts of lead. It acts as a chelating agent for heavy metal poisoning. As a ligand, it forms complexes with metals like zinc, lead and mercury. Further, it is used to assess the purity of human pancreatic islet preparations, which is used for transplantation in patients with type 1 diabetes. In addition to this, it is used as a titrant in the quantitative determination of heavy metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-10-6
Molecular Formula C13H12N4S
Molecular Weight (g/mol) 256.33
MDL Number MFCD00003025
InChI Key UOFGSWVZMUXXIY-UHFFFAOYSA-N
Synonym dithizone, dithizon, ditizon, carbazone, diphenylthio, usaf ek-3092, diazenecarbothioic acid, phenyl-, 2-phenylhydrazide, 1,5-diphenyl-3-mercaptoformazan, 3-formazanthiol, 1,5-diphenyl, unii-njz2cj4d6p, 1,5-diphenyl-3-thiocarbazone
PubChem CID 657262
IUPAC Name 1-anilino-3-phenyliminothiourea
SMILES S=C(NNC1=CC=CC=C1)N=NC1=CC=CC=C1

Specifications

Melting Point 168°C (decomposition)
Odor Odorless
Quantity 2 g
Beilstein 748838
Merck Index 14,3377
Solubility Information Soluble in ethanol,chloroform,dimethyl sulfoxide and pyridine. Insoluble in water.
Formula Weight 256.33
Percent Purity ≥85%
Grade ACS
Physical Form Crystalline
Chemical Name or Material Dithizone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'