missing translation for 'onlineSavingsMsg'
Learn More

Disulfiram, TRC

Product Code. 30134688
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134688 50 g 50mg
30134060 100 g 100g
18090620 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134688 Supplier Toronto Research Chemicals Supplier No. TRCD49364050MG
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-77-8
Molecular Formula C10 H20 N2 S4
Molecular Weight (g/mol) 296.54
Synonym Disulfiram, tetraethyldisulfanedicarbothioamide, Thioperoxydicarbonic diamide ([(H2N)C(S)]2S2), N,N,N',N'-tetraethyl-, Disulfide, bis(diethylthiocarbamoyl) (8CI), Thioperoxydicarbonic diamide ([(H2N)C(S)]2S2), tetraethyl- (9CI), Abstensil, Abstinil, Abstinyl, Accel TET, Accel TET-R, Akrochem TETD, Alcophobin, Antabus, Antabuse, Antadix, Antaethyl, Antalcol, Antetan, Antetil, Anticol, Antietanol, Antietil, Antikol, Antivitium, Aversan, Averzan, Bis(N,N-diethylthiocarbamoyl) disulfide, Bis(diethylthiocarbamoyl) disulfide, Contralin, Cronetal, Curekind TETD, Dicupral, Disulfiram, Ekagom DTET, Ekagom TEDS, Ekagom TETDS, Espenal, Esperal, Etabus, Ethyl Thiram, Ethyl Thiurad, Ethyl Tuads, Ethyl Tuex, Etiltox, Exhoran, Exhorran, Hoca, Krotenal, N,N,N',N'-Tetraethyldithiuram disulfide, N,N,N',N'-Tetraethylthiuram disulfide, NSC 25953, Nocceler TED, Nocceler TET, Nocceler TET-G, Noxal, Refusal, Sanceler TET, Sanceler TET-G, Soxinol TET, Stopetyl, TETD, TTD, TTs, Tetradin, Tetradine, Tetraethyldithiuram disulfide, Tetraethylthioperoxydicarbonic diamide, Tetraethylthiram disulfide, Tetraethylthiuram, Tetraethylthiuram disulfide, Tetraethylthiuram sulfide, Tetraetil, Teturam, Teturamin, Thiuram E, Thiuranide
IUPAC Name diethylcarbamothioylsulfanyl N,N-diethylcarbamodithioate
SMILES CCN(CC)C(=S)SSC(=S)N(CC)CC

Specifications

Quantity 50 g
InChI Formula InChI=1S/C10H20N2S4/c1-5-11(6-2)9(13)15-16-10(14)12(7-3)8-4/h5-8H2,1-4H3
Formula Weight 296.0509
Chemical Name or Material Disulfiram
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.