missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Disulfiram

Product Code. 18792714
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18792714 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18792714 Supplier Tocris Bioscience™ Supplier No. 3807/50
  • ONLINE EXCLUSIVE PRICE
    £98.00
    £92.40 / 50mg
    Save £5.60
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

Reversibly stimulates SERCA Ca2+-ATPase; displays a range of other activities

Inhibitor of aldehyde dehydrogenase that displays antialcoholism activity. Shown to reversibly stimulate Ca2+-ATPase activity and inhibit V-ATPase (EC50 = 26 μM). Also inhibits expression of MMP-2 and MMP-9 and displays anti-invasive activity. Pyroptosis inhibitor; binds gasdermin D (GSDMD) and inhibits pore formation. Also inhibits SARS-CoV-2 main protease (Mpro; IC50 = 9.35 μM) and inhibits viral replication in Vero cells in vitro.
TRUSTED_SUSTAINABILITY

Specifications

CAS 97-77-8
Quantity 50 mg
Molecular Formula C10H20N2S4
Formula Weight 296.54
Purity >98%
Molecular Weight (g/mol) 296.524
Chemical Name or Material Disulfiram
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.