missing translation for 'onlineSavingsMsg'
Learn More

Disulfiram, Mikromol™

Product Code. 30051633
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051633 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051633 Supplier Mikromol Supplier No. MM1946.000250
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Disulfiram
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Disulfiram
CAS 97-77-8
Analyte or Component Names Disulfiram
Shipping Condition Room Temperature
Molecular Formula C10 H20 N2 S4
Synonym Disulfiram, tetraethyldisulfanedicarbothioamide, Thioperoxydicarbonic diamide ([(H2N)C(S)]2S2), N,N,N',N'-tetraethyl-, Disulfide, bis(diethylthiocarbamoyl) (8CI), Thioperoxydicarbonic diamide ([(H2N)C(S)]2S2), tetraethyl- (9CI), Abstensil, Abstinil, Abstinyl, Accel TET, Accel TET-R, Akrochem TETD, Alcophobin, Antabus, Antabuse, Antadix, Antaethyl, Antalcol, Antetan, Antetil, Anticol, Antietanol, Antietil, Antikol, Antivitium, Aversan, Averzan, Bis(N,N-diethylthiocarbamoyl) disulfide, Bis(diethylthiocarbamoyl) disulfide, Contralin, Cronetal, Curekind TETD, Dicupral, Disulfiram, Ekagom DTET, Ekagom TEDS, Ekagom TETDS, Espenal, Esperal, Etabus, Ethyl Thiram, Ethyl Thiurad, Ethyl Tuads, Ethyl Tuex, Etiltox, Exhoran, Exhorran, Hoca, Krotenal, N,N,N',N'-Tetraethyldithiuram disulfide, N,N,N',N'-Tetraethylthiuram disulfide, NSC 25953, Nocceler TED, Nocceler TET, Nocceler TET-G, Noxal, Refusal, Sanceler TET, Sanceler TET-G, Soxinol TET, Stopetyl, TETD, TTD, TTs, Tetradin, Tetradine, Tetraethyldithiuram disulfide, Tetraethylthioperoxydicarbonic diamide, Tetraethylthiram disulfide, Tetraethylthiuram, Tetraethylthiuram disulfide, Tetraethylthiuram sulfide, Tetraetil, Teturam, Teturamin, Thiuram E, Thiuranide
InChI Formula InChI=1S/C10H20N2S4/c1-5-11(6-2)9(13)15-16-10(14)12(7-3)8-4/h5-8H2,1-4H3
SMILES CCN(CC)C(=S)SSC(=S)N(CC)CC
IUPAC Name diethylcarbamothioylsulfanyl N,N-diethylcarbamodithioate
Molecular Weight (g/mol) 296.54
Quantity 250 mg
Formula Weight 296.0509 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.