missing translation for 'onlineSavingsMsg'
Learn More

Disulfiram, MedChemExpress

Product Code. 16490838
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16490838 5 g 5g
16442109 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16490838 Supplier Medchem Express Supplier No. HYB0240
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Disulfiram (Tetraethylthiuram disulfide) is a specific inhibitor of aldehyde-dehydrogenase (ALDH1), used for the treatment of chronic alcoholism by producing an acute sensitivity to alcohol.

Disulfiram inhibits gasdermin D (GSDMD) pore formation in liposomes and inflammasome-mediated pyroptosis and IL-1β secretion in human and mouse cells. Disulfiram + Cu2+ increases intracellular ROS levels triggering apoptosis of ovarian cancer stem cells[1-6].

Specifications

Chemical Name or Material Disulfiram
Physical Form Powder
CAS 97-77-8
Color Off-White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C10H20N2S4
For Use With (Application) Cancer-programmed cell death
Synonym Tetraethylthiuram disulfide TETD
Solubility Information DMSO : 75 mg/mL (252.92 mM; Need ultrasonic)
SMILES S=C(SSC(N(CC)CC)=S)N(CC)CC
Molecular Weight (g/mol) 296.54
Formula Weight 296.54
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.