missing translation for 'onlineSavingsMsg'
Learn More

Disuccinimidyl glutarate, 97%

Product Code. 15414538
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15414538 500 mg 500mg
15404538 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15414538 Supplier Thermo Scientific Alfa Aesar Supplier No. H58208.ME
  • ONLINE EXCLUSIVE PRICE
    £431.46
    £353.65 / 500mg
    Save £77.81
    18% Off
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 79642-50-5 | C13H14N2O8 | 326.26 g/mol

Disuccinimidyl glutarate is used as a membrane-soluble, amine-reactive N-hydroxysuccinamide ester-based homobifunctional crosslinking reagent. It is a homobifunctionla cross-linking reagent, for ex. isolation of the insulin receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Disuccinimidyl glutarate is used as a membrane-soluble, amine-reactive N-hydroxysuccinamide ester-based homobifunctional crosslinking reagent. It is a homobifunctionla cross-linking reagent, for ex. isolation of the insulin receptor.

Solubility
Soluble in dimethylsulfoxide and dimethylformamide. Partially soluble in water.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79642-50-5
Molecular Formula C13H14N2O8
Molecular Weight (g/mol) 326.26
MDL Number MFCD00153597
InChI Key LNQHREYHFRFJAU-UHFFFAOYSA-N
Synonym disuccinimidyl glutarate, di n-succinimidyl glutarate, bis 2,5-dioxopyrrolidin-1-yl pentanedioate, pentanedioic acid,1,5-bis 2,5-dioxo-1-pyrrolidinyl ester, dsg crosslinker, glutaric acid disuccinimidyl ester, di n-hydroxysuccinimidyl glutarate, bis 2,5-dioxopyrrolidin-1-yl glutarate, di n-succinimidyl glutarate chn
PubChem CID 4432628
IUPAC Name bis(2,5-dioxopyrrolidin-1-yl) pentanedioate
SMILES O=C(CCCC(=O)ON1C(=O)CCC1=O)ON1C(=O)CCC1=O

Specifications

Quantity 500 mg
Beilstein 8575145
Sensitivity Moisture sensitive
Solubility Information Soluble in dimethylsulfoxide and dimethylformamide. Partially soluble in water.
Formula Weight 326.26
Percent Purity 97%
Chemical Name or Material Disuccinimidyl glutarate
compliance-icons
Product Identifier
  • Disuccinimidyl glutarate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Pentanedioic acid, 1,5-bis(2,5-dioxo-1-pyrrolidinyl) ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.