Learn More
Lenz Laborglasintrumente Distilling Receivers Acc.to Bernauer with Flasks
Click to view available options
Standard Taper Joint:
Upper Socket 12/23 NS
Upper Socket 19/26 NS
Unit Size:
Each
Description
- Distilling receivers acc. to Bernauer (modified)
- With three cones NS 14/23 and three flasks
- Vacuum connection with glass olive
- Made of DURAN™ tubing
Specifications
Specifications
| Material | Duran Glass |
| Capacity (Metric) | 100 mL |
| Quantity | 1 Ea. |
| Standard Taper Joint | Upper Socket 19/26 NS |